accessibility menu, dialog, popup

UserName

CAS RN 6068-76-4

IUPAC Name: 3-hydroxy-2-(2-hydroxyphenyl)-4H-chromen-4-one
Synonyms: 3-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: VECGDSZOFMYGAF-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=C(O)C(=O)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results