accessibility menu, dialog, popup

UserName

OOOOOO

CAS RN 6053-46-9

IUPAC Name: 4,10-dioxatricyclo[6.3.1.0²,⁷]dodecane-3,5,9,11-tetrone
Synonyms: 4,10-dioxatricyclo[6.3.1.0²,⁷]dodecane-3,5,9,11-tetrone
Molecular Weight (g/mol): 224.17
Molecular Formula: C10H8O6
InChi Key: QVEIRZNRYOJFCL-UHFFFAOYNA-N
SMILES: O=C1CC2C3CC(C2C(=O)O1)C(=O)OC3=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results