accessibility menu, dialog, popup

UserName

H2OH2OFeOOOO

CAS RN 6047-25-2

IUPAC Name: λ²-iron(2+) dihydrate oxalate
Synonyms: ferrous oxalate dihydrate
Molecular Weight (g/mol): 179.89
Molecular Formula: C2H4FeO6
InChi Key: NPLZZSLZTJVZSX-UHFFFAOYSA-L
SMILES: O.O.[Fe++].[O-]C(=O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)