accessibility menu, dialog, popup

UserName

CAS RN 6033-87-0

NK

CAS RN 6033-87-0

IUPAC Name: potassium 9H-carbazol-9-ide
Synonyms: potassium 9H-carbazol-9-ide
Molecular Weight (g/mol): 205.3
Molecular Formula: C12H8KN
InChi Key: WTSKDGZCVMJGDJ-UHFFFAOYSA-N
SMILES: [K+].[N-]1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results