accessibility menu, dialog, popup

UserName

Sb

CAS RN 603-36-1

IUPAC Name: triphenylstibane
Synonyms: triphenylstibine
Molecular Weight (g/mol): 353.08
Molecular Formula: C18H15Sb
InChi Key: HVYVMSPIJIWUNA-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Sb](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results