CAS RN 6009-50-3
IUPAC Name:
N-{4-[2-methyl-4-(phenylamino)-1,2,3,4-tetrahydroquinoline-1-carbonyl]phenyl}methanesulfonamide
Synonyms:
N-{4-[2-methyl-4-(phenylamino)-3,4-dihydro-2H-quinoline-1-carbonyl]phenyl}methanesulfonamide
Molecular Weight (g/mol):
435.54
Molecular Formula:
C24H25N3O3S
InChi Key:
KCIJVZDZECUOJA-UHFFFAOYNA-N
SMILES:
CC1CC(NC2=CC=CC=C2)C2=CC=CC=C2N1C(=O)C1=CC=C(NS(C)(=O)=O)C=C1
1
–
1
of
1
results
1
Promotions Available
Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane, 97%
Bis(4-methoxyphenyl)-1,1,2,2-tetramethyldisilane, 97%
CAS: 6009-50-3 Molecular Formula: C24H25N3O3S Molecular Weight (g/mol): 435.54 MDL Number: MFCD09266190 InChI Key: KCIJVZDZECUOJA-UHFFFAOYNA-N Synonym: bis 4-methoxyphenyl-1,1,2,2-tetramethyldisilane,1,2-bis 4-methoxyphenyl-1,1,2,2-tetramethyldisilane,amtsi087,1,1,2,2-tetramethyl-1,2-bis 4-methoxyphenyl disilane,disilane,1,2-bis 4-methoxyphenyl-1,1,2,2-tetramethyl PubChem CID: 14866857 IUPAC Name: (4-methoxyphenyl)-[(4-methoxyphenyl)-dimethylsilyl]-dimethylsilane SMILES: CC1CC(NC2=CC=CC=C2)C2=CC=CC=C2N1C(=O)C1=CC=C(NS(C)(=O)=O)C=C1