accessibility menu, dialog, popup

UserName

CAS RN 599-00-8

IUPAC Name: trifluoroethan(²H)oic acid
Synonyms: trifluoroethan(²H)oic acid
Molecular Weight (g/mol): 115.03
Molecular Formula: C2HF3O2
InChi Key: DTQVDTLACAAQTR-DYCDLGHISA-N
SMILES: [2H]OC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results