accessibility menu, dialog, popup

UserName

CAS RN 59886-90-7

CH3CH3HCNHOCl

CAS RN 59886-90-7

IUPAC Name: 4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Synonyms: 4-chloro-5,6-dimethyl-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Molecular Weight (g/mol): 158.61
Molecular Formula: C7H9ClNO
InChi Key: HHYDNQMGLBHVSW-UHFFFAOYNA-N
SMILES: CC1=C(C)C(Cl)=C[CH-][NH++]1[O-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results