CAS RN 597-64-8
IUPAC Name:
tetraethylstannane
Synonyms:
tetraethyltin tetraethyl tin
Molecular Weight (g/mol):
234.96
Molecular Formula:
C8H20Sn
InChi Key:
RWWNQEOPUOCKGR-UHFFFAOYSA-N
SMILES:
CC[Sn](CC)(CC)CC
1
–
1
of
1
results
1
Tetraethyltin, 98%
CAS: 597-64-8 Molecular Formula: C8H20Sn Molecular Weight (g/mol): 234.96 MDL Number: MFCD00009020 InChI Key: RWWNQEOPUOCKGR-UHFFFAOYSA-N Synonym: tetraethyltin,stannane, tetraethyl,tetraethyl tin,tin, tetraethyl,unii-31re6na75o,et4sn,snet4,c2h5 4sn,rwwnqeopuockgr-uhfffaoysa PubChem CID: 11704 IUPAC Name: tetraethylstannane SMILES: CC[Sn](CC)(CC)CC