accessibility menu, dialog, popup

UserName

CAS RN 5963-14-4

IUPAC Name: 8-methylnonanoate
Synonyms: 8-methylnonanoate
Molecular Weight (g/mol): 171.26
Molecular Formula: C10H19O2
InChi Key: OAOABCKPVCUNKO-UHFFFAOYSA-M
SMILES: CC(C)CCCCCCC([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)