accessibility menu, dialog, popup

UserName

HClBiHCl

CAS RN 594-30-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 513.21
Molecular Formula: C18H17BiCl2
InChi Key: HNWIOYSWSXFTEX-UHFFFAOYSA-N
SMILES: Cl.Cl.C1=CC=C(C=C1)[Bi](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results