accessibility menu, dialog, popup

UserName

CAS RN 5928-51-8

IUPAC Name: 3-(thiophen-2-yl)propanoate
Synonyms: 3-(thiophen-2-yl)propanoate
Molecular Weight (g/mol): 155.19
Molecular Formula: C7H7O2S
InChi Key: MJPVYTKZYZPIQA-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCC1=CC=CS1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results