accessibility menu, dialog, popup

UserName

CH3CH3(E)

CAS RN 592-47-2

IUPAC Name: (3E)-hex-3-ene
Synonyms: trans-3-hexene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: ZQDPJFUHLCOCRG-AATRIKPKSA-N
SMILES: CC\C=C\CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results