CAS RN 592-35-8
CAS RN 592-35-8
IUPAC Name:
butyl carbamate
Synonyms:
O-butyl carbamate
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
SKKTUOZKZKCGTB-UHFFFAOYSA-N
SMILES:
CCCCOC(N)=O
1
–
1
of
1
results
1
Butyl Carbamate 98.0+%, TCI America™
CAS: 592-35-8 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.148 MDL Number: MFCD00007967 InChI Key: SKKTUOZKZKCGTB-UHFFFAOYSA-N Synonym: n-butyl carbamate,carbamic acid, butyl ester,usaf fo-1,usaf el-101,carbamic acid butyl ester,unii-my8i82ajs5,ccris 5069,my8i82ajs5,butyl aminooate,o-butyl carbamate PubChem CID: 11596 IUPAC Name: butyl carbamate SMILES: CCCCOC(=O)N