accessibility menu, dialog, popup

UserName

CAS RN 591-68-4

H3COOCH3

CAS RN 591-68-4

IUPAC Name: butyl pentanoate
Synonyms: pentanoic acid, butyl ester
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: OKJADYKTJJGKDX-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results