accessibility menu, dialog, popup

UserName

CAS RN 591-23-1

CH3HO

CAS RN 591-23-1

IUPAC Name: 3-methylcyclohexan-1-ol
Synonyms: 3-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: HTSABYAWKQAHBT-UHFFFAOYNA-N
SMILES: CC1CCCC(O)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results