CAS RN 590417-55-3
IUPAC Name:
4-bromo-1-methyl-1H-indole
Synonyms:
4-bromo-1-methylindole
Molecular Weight (g/mol):
210.07
Molecular Formula:
C9H8BrN
InChi Key:
JZOSXTYDJPHXQD-UHFFFAOYSA-N
SMILES:
CN1C=CC2=C(Br)C=CC=C12
1
–
1
of
1
results
1
4-Bromo-1-methyl-1H-indole, ≥97%, Thermo Scientific™
CAS: 590417-55-3 Molecular Formula: C9H8BrN Molecular Weight (g/mol): 210.07 MDL Number: MFCD08435875 InChI Key: JZOSXTYDJPHXQD-UHFFFAOYSA-N Synonym: 4-bromo-1-methyl-1h-indole,4-bromo-1-methyl-indole,acmc-209m9m,1h-indole,4-bromo-1-methyl,4-bromo-1-methylindole,1h-indole, 4-bromo-1-methyl PubChem CID: 22558937 IUPAC Name: 4-bromo-1-methylindole SMILES: CN1C=CC2=C(Br)C=CC=C12