CAS RN 58960-00-2
IUPAC Name:
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
Synonyms:
2-methyl-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol):
165.25
Molecular Formula:
C9H11NS
InChi Key:
QZSCUJZRZIIVBM-UHFFFAOYNA-N
SMILES:
CC1CNC2=CC=CC=C2S1
1
–
1
of
1
results
1
Promotions Available
2-Methyl-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
2-Methyl-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
CAS: 58960-00-2 Molecular Formula: C9H11NS Molecular Weight (g/mol): 165.254 MDL Number: MFCD12027245 InChI Key: QZSCUJZRZIIVBM-UHFFFAOYSA-N Synonym: 2h-1,4-benzothiazine, 3,4-dihydro-2-methyl,3,4-dihydro-2-methyl-2h-1,4-benzothiazine,2-methyl-3,4-dihydro-2h-benzo b 1,4 thiazine PubChem CID: 13183653 IUPAC Name: 2-methyl-3,4-dihydro-2H-1,4-benzothiazine SMILES: CC1CNC2=CC=CC=C2S1