CAS RN 588-07-8
IUPAC Name:
N-(3-chlorophenyl)acetamide
Synonyms:
3-chloroacetanilide
Molecular Weight (g/mol):
169.61
Molecular Formula:
C8H8ClNO
InChi Key:
MUUQHCOAOLLHIL-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=CC(Cl)=C1
1
–
1
of
1
results
1
3'-Chloroacetanilide 98.0+%, TCI America™
CAS: 588-07-8 Molecular Formula: C8H8ClNO Molecular Weight (g/mol): 169.608 MDL Number: MFCD00016341 InChI Key: MUUQHCOAOLLHIL-UHFFFAOYSA-N Synonym: 3'-chloroacetanilide,3-chloroacetanilide,n-3-chlorophenyl acetamide,m-chloroacetanilide,acetamide, n-3-chlorophenyl,acetanilide, 3'-chloro,meta-chloroacetanilide,unii-74h09hu61j,acetic acid, amide, n 3-chlorophenyl,m-chloroacetoanilide PubChem CID: 11493 IUPAC Name: N-(3-chlorophenyl)acetamide SMILES: CC(=O)NC1=CC(=CC=C1)Cl