accessibility menu, dialog, popup

UserName

CAS RN 58794-09-5

NBr

CAS RN 58794-09-5

IUPAC Name: 7-bromoisoquinoline
Synonyms: 7-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: KABRXLINDSPGDF-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CN=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results