accessibility menu, dialog, popup

UserName

CH3OCH3O

CAS RN 5878-19-3

IUPAC Name: 1-methoxypropan-2-one
Synonyms: 1-methoxy-2-propanone
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: CUZLJOLBIRPEFB-UHFFFAOYSA-N
SMILES: COCC(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)