accessibility menu, dialog, popup

UserName

NHNSOOCl

CAS RN 58767-51-4

IUPAC Name: 1H-imidazole-5-sulfonyl chloride
Synonyms: 3H-imidazole-4-sulfonyl chloride
Molecular Weight (g/mol): 166.58
Molecular Formula: C3H3ClN2O2S
InChi Key: MXCZCMCHRWGSBX-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CN=CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)