accessibility menu, dialog, popup

UserName

H2CCH2

CAS RN 5876-87-9

IUPAC Name: dodeca-1,11-diene
Synonyms: 1,11-dodecadiene
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: IYPLTVKTLDQUGG-UHFFFAOYSA-N
SMILES: C=CCCCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results