accessibility menu, dialog, popup

UserName

H3COH

CAS RN 5876-76-6

IUPAC Name: 4-phenylbut-3-yn-2-ol
Synonyms: 4-phenylbut-3-yn-2-ol
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: JYOZFNMFSVAZAW-UHFFFAOYNA-N
SMILES: CC(O)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)