CAS RN 5870-68-8
CAS RN 5870-68-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 3-Methylvalerate 98.0+%, TCI America™
CAS: 5870-68-8 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.214 MDL Number: MFCD00041390 InChI Key: TXAWGHYFBQBVNK-UHFFFAOYSA-N Synonym: ethyl 3-methylvalerate,pentanoic acid, 3-methyl-, ethyl ester,ethyl beta-methylvalerate,valeric acid, 3-methyl-, ethyl ester,3-methylvaleric acid ethyl ester,3-methylpentanoic acid, ethyl ester,fema no. 3679,ethyl 3-methyl valerate,ethyl 3-methyl pentanoate PubChem CID: 560255 IUPAC Name: ethyl 3-methylpentanoate SMILES: CCC(C)CC(=O)OCC