accessibility menu, dialog, popup

UserName

CH3CH3H3COOH2NHCl

CAS RN 58640-01-0

IUPAC Name: tert-butyl 4-aminobutanoate hydrochloride
Synonyms: tert-butyl 4-aminobutanoate hydrochloride
Molecular Weight (g/mol): 195.69
Molecular Formula: C8H18ClNO2
InChi Key: CZVNLCOJFFYZPG-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)OC(=O)CCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)