accessibility menu, dialog, popup

UserName

CAS RN 58586-81-5

IUPAC Name: 4-ethoxybenzohydrazide
Synonyms: 4-ethoxybenzohydrazide
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: STTILZBQLOSDNQ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results