accessibility menu, dialog, popup

UserName

CAS RN 584-12-3

BrO

CAS RN 584-12-3

IUPAC Name: 2-bromofuran
Synonyms: 2-bromofuran
Molecular Weight (g/mol): 146.97
Molecular Formula: C4H3BrO
InChi Key: OYMCMWPHMPODNK-UHFFFAOYSA-N
SMILES: BrC1=CC=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results