accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 583-57-3

IUPAC Name: 1,2-dimethylcyclohexane
Synonyms: 1,2-dimethylcyclohexane 1,2-dimethylcyclohexane (cis)
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: KVZJLSYJROEPSQ-UHFFFAOYNA-N
SMILES: CC1CCCCC1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results