accessibility menu, dialog, popup

UserName

CAS RN 58139-59-6

NH2SNSH2NN

CAS RN 58139-59-6

IUPAC Name: 4-(2-amino-1,3-thiazol-4-yl)-1,3-thiazol-2-amine
Synonyms: 4-(2-amino-1,3-thiazol-4-yl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 198.26
Molecular Formula: C6H6N4S2
InChi Key: MRFMTBTUKQIBDI-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CSC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results