accessibility menu, dialog, popup

UserName

CAS RN 58105-91-2

HNNHNHNHHNHNHClHClHClHClHClHCl

CAS RN 58105-91-2

IUPAC Name: 1,4,7,10,13,16-hexaazacyclooctadecane hexahydrochloride
Synonyms: 1,4,7,10,13,16-hexaazacyclooctadecane hexahydrochloride
Molecular Weight (g/mol): 477.16
Molecular Formula: C12H36Cl6N6
InChi Key: GYXOJYBVPYIMAW-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.Cl.Cl.C1CNCCNCCNCCNCCNCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results