CAS RN 58101-60-3
IUPAC Name:
methyl cyclopent-3-ene-1-carboxylate
Synonyms:
methyl cyclopent-3-ene-1-carboxylate
Molecular Weight (g/mol):
126.16
Molecular Formula:
C7H10O2
InChi Key:
CEOILRYKIJRPBZ-UHFFFAOYSA-N
SMILES:
COC(=O)C1CC=CC1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals
Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals
CAS: 58101-60-3 Molecular Formula: C7H10O2 Molecular Weight (g/mol): 126.16 MDL Number: MFCD04038661 InChI Key: CEOILRYKIJRPBZ-UHFFFAOYSA-N Synonym: methyl 3-cyclopentenecarboxylate,methyl cyclopent-3-enecarboxylate,methyl-3-cyclopentene-1-carboxylate,methyl3-cyclopentenecarboxylate,3-cyclopentene-1-carboxylic acid methyl ester,3-cyclopentene-1-carboxylic acid, methyl ester,acmc-209m2j,methyl cyclopent-3-ene carboxylate,4-methoxycarbonyl cyclopent-1-ene,methyl 3-cyclopentene-1-carboxylate PubChem CID: 1514118 IUPAC Name: methyl cyclopent-3-ene-1-carboxylate SMILES: COC(=O)C1CC=CC1