accessibility menu, dialog, popup

UserName

CH3OOCH3OCH3

CAS RN 5803-30-5

IUPAC Name: 1-(2,5-dimethoxyphenyl)propan-1-one
Synonyms: 1-(2,5-dimethoxyphenyl)propan-1-one
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: DKVCHMQHUMLKPO-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(OC)=CC=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results