accessibility menu, dialog, popup

UserName

CH3CH3ClO

CAS RN 57526-28-0

IUPAC Name: 2-methylbutanoyl chloride
Synonyms: 2-methylbutanoyl chloride
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: XRPVXVRWIDOORM-UHFFFAOYNA-N
SMILES: CCC(C)C(Cl)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)