accessibility menu, dialog, popup

UserName

CH3OH3C

CAS RN 5748-38-9

IUPAC Name: 1-{4'-methyl-[1,1'-biphenyl]-4-yl}ethan-1-one
Synonyms: 1-{4'-methyl-[1,1'-biphenyl]-4-yl}ethanone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: GNIQQKORSMFYPE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results