CAS RN 57265-65-3
IUPAC Name:
1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-2,3-dihydro-1H-imidazole hydrochloridyl
Synonyms:
1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-2H-imidazole hydrochloridyl
Molecular Weight (g/mol):
688.7
Molecular Formula:
C31H24Cl7N2O
InChi Key:
LBFGPQFQXRJXDI-UHFFFAOYNA-N
SMILES:
[Cl].ClC1=CC=C(C=C1)C(N1CN(CC(OCC2=CC=C(Cl)C=C2Cl)C2=CC=C(Cl)C=C2Cl)C=C1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Calmidazolium Chloride
CAS: 57265-65-3 Molecular Formula: C31H23Cl7N2O