accessibility menu, dialog, popup

UserName

CAS RN 57012-20-1

IUPAC Name: (1,3-dimethyl-1H-pyrazol-5-yl)methanol
Synonyms: (2,5-dimethylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: GUJDKMVLHCJODO-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results