accessibility menu, dialog, popup

UserName

H3CONHNH2

CAS RN 5699-41-2

IUPAC Name: N-(4-aminobutyl)acetamide
Synonyms: acetylputrescine
Molecular Weight (g/mol): 130.19
Molecular Formula: C6H14N2O
InChi Key: KLZGKIDSEJWEDW-UHFFFAOYSA-N
SMILES: CC(=O)NCCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results