accessibility menu, dialog, popup

UserName

CAS RN 569650-64-2

IUPAC Name: λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2S,5S)-1-{2-[(2S,5S)-2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane tetrafluoroboranuide
Synonyms: λ¹-rhodium(1+) (2S,5S)-1-{2-[(2S,5S)-2,5-diisopropylphospholan-1-yl]phenyl}-2,5-diisopropylphospholane 1,5-cyclooctadiene, (Z,Z)- tetrafluoroborate
Molecular Weight (g/mol): 716.48
Molecular Formula: C34H56BF4P2Rh
InChi Key: AXLKBYDAACXFBH-KFORCWLUNA-N
SMILES: [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC(C)[C@@H]1CC[C@@H](C(C)C)P1C1=CC=CC=C1P1[C@@H](CC[C@H]1C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results