CAS RN 5683-41-0
IUPAC Name:
2,5,6-trimethyl-1,3-benzothiazole
Synonyms:
2,5,6-trimethyl-1,3-benzothiazole
Molecular Weight (g/mol):
177.27
Molecular Formula:
C10H11NS
InChi Key:
ZZLZXZFVVOXVRG-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC(C)=C(C)C=C2S1
1
–
1
of
1
results
1
2,5,6-Trimethylbenzothiazole, TCI America™
CAS: 5683-41-0 Molecular Formula: C10H11NS Molecular Weight (g/mol): 177.265 MDL Number: MFCD00128832 InChI Key: ZZLZXZFVVOXVRG-UHFFFAOYSA-N Synonym: 2,5,6-trimethylbenzothiazole,2,5,6-trimethylbenzo d thiazole,benzothiazole, 2,5,6-trimethyl,pubchem20514,acmc-1b075,benzothiazole,2,5,6-trimethyl,zzlzxzfvvoxvrg-uhfffaoysa,benzothiazole, 2,5,6-trimethyl-6ci,7ci,8ci,9ci PubChem CID: 79765 IUPAC Name: 2,5,6-trimethyl-1,3-benzothiazole SMILES: CC1=CC2=C(C=C1C)SC(=N2)C