accessibility menu, dialog, popup

UserName

CAS RN 5680-61-5

H3COONOOOOCH3

CAS RN 5680-61-5

IUPAC Name: 1,3-diethyl 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanedioate
Synonyms: 1,3-diethyl 2-(1,3-dioxoisoindol-2-yl)propanedioate
Molecular Weight (g/mol): 305.29
Molecular Formula: C15H15NO6
InChi Key: SZNGBHWKVWEBKW-UHFFFAOYSA-N
SMILES: CCOC(=O)C(N1C(=O)C2=CC=CC=C2C1=O)C(=O)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results