accessibility menu, dialog, popup

UserName

CH3OO(S)NH2(S)CH3H3CHCl

CAS RN 56782-52-6

IUPAC Name: ethyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Synonyms: ethyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular Weight (g/mol): 195.69
Molecular Formula: C8H18ClNO2
InChi Key: QQGRWNMNWONMOO-LEUCUCNGSA-N
SMILES: Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)