accessibility menu, dialog, popup

UserName

NH3(R)OONOO

CAS RN 5678-48-8

IUPAC Name: (3R)-3-azaniumyl-3-(2-nitrophenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(2-nitrophenyl)propanoate
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: XXBOYULKNZTOMN-SSDOTTSWSA-N
SMILES: [NH3+][C@H](CC([O-])=O)C1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)