accessibility menu, dialog, popup

UserName

H2NH2N

CAS RN 56602-77-8

IUPAC Name: 1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine
Synonyms: 1-[3-(aminomethyl)bicyclo[2.2.1]heptan-2-yl]methanamine
Molecular Weight (g/mol): 154.26
Molecular Formula: C9H18N2
InChi Key: RPYFJVIASOJLJS-UHFFFAOYNA-N
SMILES: NCC1C2CCC(C2)C1CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results