accessibility menu, dialog, popup

UserName

CAS RN 56070-16-7

CH3OPSOCH3SSOOCH3CH3H3C

CAS RN 56070-16-7

IUPAC Name: O,O-diethyl {[(2-methylpropane-2-sulfonyl)methyl]sulfanyl}phosphonothioate
Synonyms: terbufos sulfone
Molecular Weight (g/mol): 320.42
Molecular Formula: C9H21O4PS3
InChi Key: DWZSTEUTHNUVQD-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCS(=O)(=O)C(C)(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results