CAS RN 55764-23-3
IUPAC Name:
2,5-dimethylfuran-3-thiol
Synonyms:
2,5-dimethylfuran-3-thiol
Molecular Weight (g/mol):
128.19
Molecular Formula:
C6H8OS
InChi Key:
DBBHCZMXKBCICL-UHFFFAOYSA-N
SMILES:
CC1=CC(S)=C(C)O1
1
–
1
of
1
results
1
2,5-Dimethyl-3-furanthiol 97.0+%, TCI America™
CAS: 55764-23-3 Molecular Formula: C6H8OS Molecular Weight (g/mol): 128.19 MDL Number: MFCD00801026 InChI Key: DBBHCZMXKBCICL-UHFFFAOYSA-N Synonym: 2,5-dimethyl-3-furanthiol,3-mercapto-2,5-dimethylfuran,3-furanthiol, 2,5-dimethyl,unii-wct2259x5l,2,5-dimethyl-3-mercaptofuran,fema no. 3451,2,5-dimethyl-3-furylmercaptan,2,5-dimethyl-3-furan thiol,2,5-dimethyl-furan-3-thiol,acmc-1amqa PubChem CID: 41569 IUPAC Name: 2,5-dimethylfuran-3-thiol SMILES: CC1=CC(S)=C(C)O1