accessibility menu, dialog, popup

UserName

CAS RN 55755-17-4

CH3H3N

CAS RN 55755-17-4

IUPAC Name: 2-(3-methylphenyl)ethan-1-aminium
Synonyms: 3-methylphenethylamine
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: GUERDLPJJJMIEU-UHFFFAOYSA-O
SMILES: CC1=CC=CC(CC[NH3+])=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results