accessibility menu, dialog, popup

UserName

CH3SN

CAS RN 55406-13-8

IUPAC Name: 3-methylthiophene-2-carbonitrile
Synonyms: 3-methylthiophene-2-carbonitrile
Molecular Weight (g/mol): 123.17
Molecular Formula: C6H5NS
InChi Key: ALZHYEITUZEZMT-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)