accessibility menu, dialog, popup

UserName

H3CNO

CAS RN 552-70-5

IUPAC Name: 9-methyl-9-azabicyclo[3.3.1]nonan-3-one
Synonyms: pseudopelletierine granatonine pseudopelletierin pseudopunicine granatan-3-one psi-pelletierine 9-methyl-3-granatanone pseudopelletrierin 9-methyl-3-granataninone 3-granataninone, 9-methyl-
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: RHWSKVCZXBAWLZ-UHFFFAOYNA-N
SMILES: CN1C2CCCC1CC(=O)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results