accessibility menu, dialog, popup

UserName

CAS RN 550378-78-4

NN

CAS RN 550378-78-4

IUPAC Name: 9-[3-(9H-carbazol-9-yl)phenyl]-9H-carbazole
Synonyms: 9-[3-(carbazol-9-yl)phenyl]carbazole
Molecular Weight (g/mol): 408.5
Molecular Formula: C30H20N2
InChi Key: MZYDBGLUVPLRKR-UHFFFAOYSA-N
SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results